Learn More
Bachem Fmoc-Trp(Me)-OH
SDP
Supplier:  Bachem 4052226.0001
Encompass_Preferred
Fmoc-Trp(Me)-OH, 1334509-86-2, C27H24N2O4, Mr 440.50, 1g. Fmoc-Trp(Me) was employed in the Fmoc-SPPS of compstatin analogs.
Catalog No. 50-258-807