Learn More
Apexbio Technology LLC Fmoc-Trp-OH 35737-15-6 100g
SDP
Supplier:  Apexbio Technology LLC A7977100000
Encompass_Preferred
Fmoc-Trp-OH is an Fmoc-protected tryptophan derivative frequently utilized in solid-phase peptide synthesis (SPPS) The Fmoc group temporarily protects the amino functionality of tryptophan allowing controlled peptide chain elongation and minimizing undesired side reactions during synthesis Following each coupling cycle the Fmoc group can be efficiently removed under mild basic conditions Fmoc-Trp-OH serves as a fundamental building block for synthesizing bioactive peptides peptide-derived therapeutics and protein segments Based on these properties Fmoc-Trp-OH is widely applied in biomedical research and pharmaceutical development
Catalog No. 50-000-92554