Learn More
Medchemexpress LLC Fmoc-Tyr(SO3nP)-OH | 878408-63-0 | MFCD00011361 | >=95.0% | 553.62 g/mol | C29H31NO8S | 50 MG

Supplier: Medchemexpress LLC HYW14200850MG
Fmoc-Tyr(SO3nP)-OH is an Fmoc-protected tyrosine derivative with a sulfonate-protected side chain, supplied as a solid for use in peptide synthesis and related research applications. The material is intended for research use only and should be stored sealed at low temperature to maintain stability.
- Fmoc-protected tyrosine derivative for peptide synthesis.
- Sulfonate-protected (SO3nP) side chain to mimic sulfonation.
- Molecular formula C29H31NO8S.
- Molecular weight 553.62 g/mol.
- CAS number 878408-63-0.
- Available in small quantities including 50 MG.
- Purity ≥95.0% (HPLC) as reported by suppliers.
- Store sealed at -20°C; stock solution guidance: -80°C up to 6 months, -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.