Learn More
Medchemexpress LLC Fmoc-val-ala-aminomethyl acetate | 2505045-86-1 | 97.3% | 481.54 | C26H31N3O6 | 1 G

Supplier: Medchemexpress LLC HY1568131G
Fmoc-Val-Ala-aminomethyl acetate is a valine-alanine protease-cleavable linker reagent used in the synthesis of antibody-drug conjugates (ADCs) for research applications. It is characterized by LCMS-reported purity and is used to introduce a cleavable spacer between antibody and payload in linker-drug constructs.
- Protease-cleavable valine-alanine linker.
- Suitable for ADC synthesis and linker-drug conjugation.
- Purity 97.3% (LCMS).
- Molecular weight 481.54 g/mol.
- Molecular formula C26H31N3O6.
- Supplied as a 1 G pack for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.