Learn More
Medchemexpress LLC Fmoc-Val-Ala-aminomethyl acetate | 2505045-86-1 | 97.3% | 481.54 g/mol | C26H31N3O6 | 25 MG

Supplier: Medchemexpress LLC HY15681325MG
Fmoc-Val-Ala-aminomethyl acetate is an Fmoc-protected Val-Ala dipeptide bearing an aminomethyl acetate terminus, used as an ADC linker building block in the synthesis of antibody-drug conjugates for research applications.
- CAS number: 2505045-86-1
- Chemical formula: C26H31N3O6
- Molecular weight: 481.54 g/mol
- Purity: 97.3% (LCMS)
- Available pack sizes: 25 MG and larger
- Storage: -20°C, protect from light, stored under nitrogen; in solvent: -80°C (6 months), -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.