Learn More
Medchemexpress LLC Fmoc-Val-Ala-aminomethyl acetate | 2505045-86-1 | 97.3% | 481.54 g·mol⁻¹ | C26H31N3O6 | 250 MG

Supplier: Medchemexpress LLC HY156813250MG
Fmoc-Val-Ala-aminomethyl acetate is an ADC linker used in the synthesis of antibody-drug conjugates. It provides a cleavable connection between an antibody and a cytotoxic payload for controlled release in targeted therapies.
- Purity 97.3%.
- Cas number 2505045-86-1.
- Molecular formula C26H31N3O6 and molecular weight 481.54 g·mol⁻¹.
- Soluble in DMSO at 50 mg/mL; ultrasonic treatment may be required.
- Store protected from light, under nitrogen; in solvent store at -80°C up to 6 months, -20°C up to 1 month.
- Available in small quantities suitable for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.