Learn More
Medchemexpress LLC Fmoc-Val-Ala-OH 100g
SDP
Supplier:  Medchemexpress LLC HYW106311100G
Encompass_Preferred
Fmoc-Val-Ala-OH is a reagent consisting of a Val-Ala dipeptide with an Fmoc protecting group on its N-terminus Carboxylic acids can be reactive with alcohols or amines while the Fmoc protecting group can be removed with piperidine to allow for coupling with carboxylic acids to form amides
Catalog No. 50-002-06955