Learn More
Medchemexpress LLC Fmoc-Val-Cit-PAB | 159858-22-7 | MFCD22417106 | 99.8% | 601.69 | C33H39N5O6 | 5 G
SDP
Supplier:  Medchemexpress LLC HY193185G
Encompass_Preferred
Fmoc-Val-Cit-PAB is an Fmoc-protected valine-citrulline para-aminobenzyl (PAB) cleavable linker used in antibody-drug conjugate (ADC) synthesis. It provides a protease-sensitive Val-Cit cleavage site and a PAB self-immolative spacer to enable enzymatic release of payloads in lysosomes. The compound is supplied as a high-purity solid with good solubility in DMSO for conjugation workflows.
  • Cleavable Val-Cit dipeptide recognized by cathepsin B
  • PAB self-immolative spacer for rapid payload release
  • Fmoc-protected amine compatible with standard peptide chemistry
  • High purity (99.8%) suitable for research conjugation
  • Soluble in DMSO (~83.33 mg/mL) with ultrasonic assistance
  • Store protected from light; stable frozen in solution (-80°C, 6 months)
Catalog No. 50-002-03659