Learn More
Medchemexpress LLC Fmoc-Val-Cit-PAB | 159858-22-7 | MFCD22417106 | 99.8% | 601.69 g/mol | C33H39N5O6 | 1 G
SDP
Supplier:  Medchemexpress LLC HY193181G
Encompass_Preferred
Fmoc-Val-Cit-PAB is a protease-cleavable peptide-para-aminobenzyl (PAB) linker used to construct antibody-drug conjugates (ADCs). The Fmoc-protected Val-Cit dipeptide is cleaved by lysosomal proteases, and the PAB spacer self-immolates to release the payload. Intended for research use only.
  • Protease-cleavable Val-Cit dipeptide with a self-immolative PAB spacer.
  • Fmoc-protected N-terminus compatible with standard peptide-coupling chemistries.
  • High purity suitable for conjugation chemistry and analytical applications.
  • Stable as a solid under recommended storage; aliquot solutions to avoid repeated freeze-thaw.
Catalog No. 50-002-03328