Learn More
Medchemexpress LLC Fmoc-val-cit-pab-duocarmycin | 99.4% | 1050.59 g·mol⁻¹ | C58H60ClN7O10 | 1 MG

Supplier: Medchemexpress LLC HY1265321MG
Fmoc-Val-Cit-PAB-Duocarmycin is a drug-linker conjugate intended for use in antibody-drug conjugate (ADC) research. It couples a Duocarmycin payload to an Fmoc-Val-Cit-PAB cleavable linker, and is supplied as a solid with reported high purity and defined storage conditions.
- High purity (99.44%).
- Molecular weight 1050.59 g·mol⁻¹.
- Molecular formula C58H60ClN7O10.
- Solid form, white to yellow.
- Storage: powder -20 °C; in solvent -80 °C for 6 months or -20 °C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.