Learn More
Medchemexpress LLC Fmoc-Val-Cit-PAB-PNP |10g
SDP
Supplier:  Medchemexpress LLC HY4118910G
Encompass_Preferred
Fmoc-Val-Cit-PAB-PNP is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) Fmoc-Val-Cit-PAB-PNP has superior plasma stability comparable to that of non-cleavable linkers[1][2][3]
Catalog No. 50-002-03584