Learn More
Medchemexpress LLC Fmoc-Val-Cit-PAB-PNP | 863971-53-3 | MFCD22200278 | 99.6% | 766.80 | C40H42N6O10 | 1 G
SDP
Supplier:  Medchemexpress LLC HY411891G
Encompass_Preferred
Fmoc-Val-Cit-PAB-PNP is a protease-cleavable antibody-drug conjugate (ADC) linker containing a valine-citrulline dipeptide and a p-aminobenzyl (PAB) self-immolative spacer; enzymatic cleavage releases carbamate-linked payloads for intracellular delivery.
  • Contains Val-Cit dipeptide and p-aminobenzyl (PAB) spacer.
  • Cleavable by lysosomal proteases such as cathepsin B.
  • Enables enzymatic release of carbamate-linked payloads.
  • Demonstrates high plasma stability compared with non-cleavable linkers.
  • Supplied with high purity (99.6%).
  • Available in milligram to gram scales for development and synthesis workflows.
Catalog No. 50-002-04084