Learn More
Apexbio Technology LLC Fmoc-Val-OH 68858-20-8 100g
SDP
Supplier:  Apexbio Technology LLC A7993100000
Encompass_Preferred
Fmoc-Val-OH (CAS 68858-20-8) is a valine derivative structurally modified at its amino terminus with a 9-fluorenylmethoxycarbonyl (Fmoc) protecting group The compound is utilized as an intermediate building block for introducing valine residues into synthetic peptides enabling the construction of peptides used in pharmaceutical research structural biology and functional analysis of bioactive sequences Based on these properties Fmoc-Val-OH holds research potential in drug design and peptide-mediated biomedical investigations
Catalog No. 50-000-88664