Learn More
Medchemexpress LLC Fmoc-VAP-MMAE | 1912408-91-3 | 1259.57 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY148339
Encompass_Preferred
Fmoc-VAP-MMAE is an agent-linker conjugate for Antibody-Drug Conjugates (ADC). It is composed of an ADC linker, a tubulin inhibitor known as MMAE, and a protective Fmoc group.
  • Agent-linker conjugate for ADC
  • Composed of an ADC linker
  • Includes a tubulin inhibitor (MMAE)
  • Features a protective Fmoc group
Catalog No. 50-003-64930