Learn More
Medchemexpress LLC Fmoc-VAP-MMAE | 1912408-91-3 | 1259.57 | 50 MG
SDP
Supplier:  Medchemexpress LLC 50MGHY148339
Encompass_Preferred
Fmoc-VAP-MMAE is an agent-linker conjugate for Antibody-Drug Conjugates (ADC). It consists of an ADC linker and a tubulin inhibitor MMAE, with a protective Fmoc group. This product is for research use only.
  • Agent-linker conjugate for ADC
  • Contains an ADC linker
  • Includes a tubulin inhibitor MMAE
  • Protected by an Fmoc group
Catalog No. 50-003-66200