Learn More
Cayman Chemical FNIII14sodIum salt 5mg
SDP
Supplier:  Cayman Chemical 413965 MG
Encompass_Preferred
A peptide inhibitor of cell adhesion; inhibits the adhesion of A375-SM cells to fibronectin-coated plates at 200 UG/ml; inhibits the binding of an antibody targeting the active conformation of β1 integrin on K562 cells and induces the differentiation of ST 13 fibroblasts into adipocytes at 100 UG/ml; decreases atherosclerotic plaque formation in Ldlr-/- mice fed a Western diet; reduces metastatic tumor burden in a 4T1 murine mammary model of metastasis in combination with doxorubicin
Catalog No. 50-294-4024