Learn More
Chemglass Life Sciences Funnel, Standard Stem, 59.1mL, 65mm Top ID
SDP
Supplier:  Chemglass Life Sciences CG177002
Encompass_Preferred
Funnel, Standard Stem, 59.1mL, 65mm Top ID. Polypropylene funnels feature the exclusive filter-step internal design for speedier filtrations. External ribs prevent airlock. The parallel stem measures 8mm OD. Steam autoclavable at 121°C (250°F).
Catalog No. 50-872-156