Learn More
Ran Biotechnologies Inc 1.5kg of FO-101 fluorinated oil
SDP
Supplier:  Ran Biotechnologies Inc FO1011500G
Encompass_Preferred
1.5kg of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)
Catalog No. NC3660288