Learn More
Bellco Glass, Inc. Foam/Level Probe
SDP
Supplier:  Bellco Glass, Inc. 780341590
Encompass_Preferred
Foam Level Probes are used in fermentation bioreactors to detect the presence of foam or the liquid level within the bioreactor vessel. When the foam level or liquid level reaches the tip of the probe, a conductance path is created for the probe to trigger the controller to react and introduce anti-foaming agent.
Catalog No. 50-194-6336