Learn More
Electron Microscopy Sciences Polyurethane Foam General Purpose Swabs with 3.3mm Round Tips
SDP
Supplier:  Electron Microscopy Sciences 7832582
Encompass_Preferred
Economical general purpose swabs. Best solvent retention. The most cost-effective swabs for general purpose applications. 100 ppi reticulated medical grade foam provides excellent particle entrapment. The open cell structure gives the best solvent holding ability of any style of swab. These swabs are free of adhesives or binders that could contaminate your process. Fiberless construction does not generate particles.
Catalog No. 50-193-0637