Learn More
Fine Science Tools Adson Forceps, Stainless Steel, Straight, 12 cm
SDP
Supplier:  Fine Science Tools 1150612
Encompass
Tip Shape: Straight/ Tips: Cross Serrated/ Tip Dimensions: 1.8 x 1.3mm/ Alloy / Material: Stainless Steel/ Length: 12cm/ Feature: Tungsten Carbide
Catalog No. NC9960996