Learn More
Bioworld Focus™ DTT, 2M, 3483-12-3, 5 vials
Supplier:  Bioworld 208400071
Encompass_Preferred
For those who prefer DTT as a reducing agent, we offer ultra-pure proteomic grade DTT for protein reduction. FOCUS™ DTT is supplied as 5 vials containing dry powder to maintain the activity of the reducing agent. Add 1ml aliquot of water to give a 2M concentration, which is then added to your buffer solution to maintain protein reduction potential. Synonym: 1,4-Dithio-DL-threitol. Molecular Weight : 154.25, Molecular Formula : C4H10O2S2, Purity : >98%, Solubility : Freely soluble in water, ethanol, acetone, ether, chloroform, and ethyl acetate. Appearance Form : Solid, Appearance Color : White to Off-White, Melting Point : 41 - 44°C, Boiling Point : 125°C, Density : 1.04 g/mL at 20°C.
Catalog No. 50-198-452