Learn More
Med Vet International Miltex Vantage Foerster Sponge Forceps, 9.5in., Curved, Serrated Jaws
Supplier:  Med Vet International MILV97610
Encompass_Preferred
The Miltex Vantage V97-610 Foerster sponge forceps are made from high quality Japanese stainless steel and measure 9.5" in length. They have a finger ring handle with a ratchet lock and curved jaws with serrations.
Catalog No. 50-282-9702