Learn More
Thomson Instrument Company Foil Heat Seal For Long Term Storage | Use w/ All Plates
SDP
Supplier:  Thomson Instrument Company 899406
Encompass_Preferred
Thomson Instrument Company’s Foil Seals are ideal for growing cultures in well plates and long-term storage. Thomson has different seals: foil seals are available with adhesive, airporous, pierceable, and sterile or non-sterile. Thomson’s well plates are offered in 24 and 96-well configurations with V, pyramid, or round bottom shapes with square or round wells for cell culture and analytical sample preparation. Well plates are designed to SLAS specifications for compatibility with automated liquid handling systems. Plates are individually wrapped and are available sterile or non-sterile for any application. Thomson additionally offers a variety of filter plates, capmats, seals and lids. Thomson manufactures its products in the USA to assure same day shipping.
Catalog No. NC2941156