Learn More
Andwin Scientific FOIL SQUARES 6IN X 6IN
SDP
Supplier:  Andwin Scientific 1086J26/DEL
Encompass
The Thomas aluminum foil is conveniently pre-cut into 6" x 6" squares. The product can be used for weighing, capping, and any other laboratory purposes. The foil squares are interleaved with paper for easy separation and are sold in a 5 lb. package.
Catalog No. NC9201519