Learn More
Selleck Chemical LLC Folic acid 100mg 59-30-3 Folacin, Vitamin B9, Vitamin M, Pteroylglutamic acid, Folate
Supplier: Selleck Chemical LLC S4605100MG
Folic acid (Folacin, Vitamin B9, Vitamin M, Pteroylglutamic acid, Folate), a B vitamin, plays an important role in cell division and in the synthesis of amino acids and nucleic acids like DNA. *For Research & Development use only. Product is not intended for drug, household, or human consumption.