Learn More
Medchemexpress LLC Folic acid-13C5 | 1207282-75-4 | 95.0% | 446.36 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY16637S31MG
Encompass_Preferred
Folic acid-13C5 is the 13C-labeled form of Folic acid. Folic acid (Vitamin M; Vitamin B9) is a B vitamin essential for the production and maintenance of new cells, as well as for DNA and RNA synthesis.
  • It can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements can be incorporated into drug molecules for quantitation during drug development.
  • Deuteration, a process related to stable isotopes, can influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-94192