Learn More
Medchemexpress LLC Folic acid | 59-30-3 | 98.4% | 441.40 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY16637R
Encompass_Preferred
Folic acid (Standard) is an analytical standard used as a reference for research and analytical applications. As Vitamin B9, it is an orally active essential nutrient from the B complex group of vitamins. It has shown antidepressant-like effects, and its sodium form can reduce the risk of neonatal neural tube defects. It is also used in research for megaloblastic and macrocytic anemias caused by folic deficiency.
  • Analytical standard for assaying
  • Used in qualitative, quantitative, and methodological research experiments
  • Commonly applied in HPLC, GC, and MS studies
  • Essential nutrient from the B complex group (Vitamin B9)
  • Shows antidepressant-like effects
  • Sodium form reduces the risk of neonatal neural tube defects
  • Used in research for megaloblastic and macrocytic anemias
Catalog No. 50-003-92311