Learn More
Medchemexpress LLC Folic acid | 59-30-3 | 99.02% | 441.40 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16637R
Encompass_Preferred
Folic acid, also known as Vitamin B9, is an orally active essential nutrient from the B complex group of vitamins. It is utilized in research for its antidepressant-like effects and its role in reducing the risk of neonatal neural tube defects. Additionally, it is studied for its involvement in megaloblastic and macrocytic anemias, prevention of chromosome breakage, and DNA hypomethylation. This compound can also be used to induce acute kidney injury models.
  • Essential nutrient from the B complex group of vitamins
  • Demonstrates antidepressant-like effects
  • Helps reduce the risk of neonatal neural tube defects (as its sodium form)
  • Utilized in research concerning megaloblastic and macrocytic anemias
  • Plays a critical role in preventing chromosome breakage
  • Involved in DNA hypomethylation research
  • Can be used to induce acute kidney injury models
Catalog No. 50-003-92661