Learn More
Cayman Chemical FolIc AcId-d4 500ug
SDP
Supplier:  Cayman Chemical 28485500 UG
Encompass_Preferred
An internal standard for the quantification of folic acid by GC- or LC-MS
Catalog No. 50-293-7798