Learn More
Medchemexpress LLC Folic acid disodium | 29704-76-5 | 98.6% | 485.36 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16637D100MG
Encompass_Preferred
Folic acid disodium is an orally active disodium salt form of Folic acid. It serves as a cofactor in single-carbon transfer reactions. It demonstrates protective effects against neural tube defects, ischemic events, and cancer. Overload can lead to impaired brain development in embryogenesis and promote the growth of precancerous altered cells, while deficiency can result in megaloblastic anemia.
  • Orally active disodium salt form of folic acid.
  • Cofactor in single-carbon transfer reactions.
  • Protects against neural tube defects, ischemic events, and cancer.
  • Overload can impair brain development and promote precancerous altered cells.
  • Deficiency can lead to megaloblastic anemia.
Catalog No. 50-003-91817