Learn More
American Radiolabeled Chemicals Inc FOLIC ACID NA SALT-3H 250UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 0125250UC
Encompass
Folic acid, sodium salt-3,5,7,9-3H 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 20-70 Ci/mmol, M.W. 441.4, Shipped in Dry Ice
Catalog No. 50-901-04272