Learn More
MOLECULAR DEPOT LLC Folic Acid Solution - 50 mL
Supplier:  MOLECULAR DEPOT LLC B2010580
Encompass_Preferred
High Folic Acid Buffered Solution is a liquid-stable buffered solution containing high levels of folic acid (20-30 ng/mL) This product has been used in the research and development of folic acid high and low working solutions (to use with Folic Acid Buffered Diluent B2010581) For in vitro research use only
Catalog No. 50-000-13536