Learn More
Medchemexpress LLC Folic acid (Standard) | 59-30-3 | 98.4% | 441.40 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16637R50MG
Encompass_Preferred
Folic acid (Standard) is the analytical standard of Folic acid, intended for research and analytical applications. Folic acid (Vitamin B9) is an orally active essential nutrient from the B complex group of vitamins. It exhibits an antidepressant-like effect. Folic acid sodium can reduce the risk of neonatal neural tube defects. Folic acid is used in the research of megaloblastic and macrocytic anemias due to folic deficiency. This compound is an analytical standard and reference standard supplied for assays.
  • Analytical standard for folic acid
  • Intended for research and analytical applications
  • Orally active essential nutrient from the B complex group of vitamins
  • Exhibits an antidepressant-like effect
  • Used to reduce the risk of neonatal neural tube defects
  • Used in research of megaloblastic and macrocytic anemias due to folic deficiency
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-003-91822