Learn More
Medchemexpress LLC Foliglurax monohydrochloride | 2133294-96-7 | 98.6% | 457.97 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY108703A50MG
Encompass_Preferred
Foliglurax monohydrochloride is a highly selective and potent, brain-penetrant metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM), with an EC50 of 79 nM. It exhibits an antiparkinsonian effect.
  • Highly selective and potent metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM)
  • Brain-penetrant
  • EC50 of 79 nM
  • Antiparkinsonian effect
Catalog No. 50-003-45128