Learn More
Medchemexpress LLC Foliglurax monohydrochloride | 2133294-96-7 | MFCD31807615 | 98.6% | 457.97 | 100 MG

Supplier: Medchemexpress LLC HY108703A100MG
Foliglurax monohydrochloride is a highly selective and potent, brain-penetrant metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM), with an EC50 of 79 nM. It exhibits an antiparkinsonian effect.
- Highly selective and potent
- Brain-penetrant
- Metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM)
- EC50 of 79 nM
- Exhibits an antiparkinsonian effect
- Chemical formula: C23H24ClN3O3S
- Appearance: Solid
- Color: Light yellow to yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.