Learn More
Selleck Chemical LLC Leucovorin Calcium Pentahydrate 50mg 6035-45-6 Folinic acid
SDP
Supplier:  Selleck Chemical LLC S123650MG
Encompass_Preferred
Leucovorin (Folinic acid) is a derivative of folic acid, which can be used to increase levels of folic acid under conditions favoring folic acid inhibition. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6133