Learn More
Medchemexpress LLC Folinic acid calcium salt pentahydrate | 6035-45-6 | 98.0% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0080500MG
Encompass_Preferred
Folinic acid calcium salt pentahydrate (Leucovorin calcium salt pentahydrate) is a biological folic acid and is generally administered along with methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity.
  • Significantly reduces micronucleated binucleated cells (MNBN) and aberrant cells (Abs).
  • Reverses growth inhibition caused by chronic methotrexate administration in mice.
Catalog No. 50-004-32567