Learn More
Medchemexpress LLC Folinic acid-d4 calcium hydrate | 58-05-9 | 91.7% | 605.61 | 250 UG
SDP
Supplier:  Medchemexpress LLC 250UGHY17556S1
Encompass_Preferred
Folinic acid-d4 calcium hydrate, also known as Leucovorin-d4 calcium hydrate, is a deuterated form of Folinic acid. It functions as a biological folic acid and is typically used as a rescue agent to reduce Methotrexate-induced toxicity. Stable heavy isotopes like deuterium are incorporated into drug molecules to serve as tracers for quantitation during drug development, which can also influence pharmacokinetic and metabolic profiles of drugs.
  • Functions as a biological folic acid
  • Reduces methotrexate-induced toxicity
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-85310