Learn More
Cusabio Technology LLC rat follicle-stimulating hormoneFSH ELISA Kit
Supplier:  Cusabio Technology LLC CSBE06869R
Encompass_Preferred
The Rat follicle-stimulating hormone (FSH) ELISA Kit is designed for the quantitative determination of endogenic rat FSH concentrations in serum, plasma, or tissue homogenates. This assay employs the quantitative sandwich enzyme immunoassay technique. Antibody specific for FSH has been pre-coated onto a microplate. Standards and samples are pipetted into the wells and any FSH present is bound by the immobilized antibody. After removing any unbound substances, a biotin-conjugated antibody specific for FSH is added to the wells. After washing, avidin conjugated Horseradish Peroxidase (HRP) is added to the wells. Following a wash to remove any unbound avidin-enzyme reagent, a substrate solution is added to the wells and color develops in proportion to the amount of FSH bound in the initial step. The color development is stopped and the intensity of the color is measured.
Catalog No. NC3654075