Learn More
ACROBiosystems Recombinant Protein;Human Follistatin-like 1 / FSTL1 Protein;HEK293;1MG;FS1,FSTL1,FRP,FSL1,FSTL-1,FSL-1
Supplier:  ACROBiosystems FS1H52271MG
Encompass_Preferred
Recombinant Protein;Human Follistatin-like 1 / FSTL1 Protein;1MG;Human FSTL1, His Tag (FS1-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 308 (Accession # AAH00055).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.7 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to differtent glycosylation.
Catalog No. 50-165-8185