Learn More
Medchemexpress LLC Fomc-gly-gly-phe-gly-oh | 1817857-75-2 | 99.66% | 558.58 | 500 MG
Supplier: Medchemexpress LLC HYP4192500MG
Fomc-Gly-Gly-Phe-Gly-OH (compound D5) can be used as an intermediate in the synthesis of ADC dual-drug-linker. This synthetic intermediate GGFGE further forms an important ADC dual-drug link assembly unit. ADCs are comprised of an antibody to which an ADC cytotoxin is attached through an ADC linker.
- Used as an intermediate in the synthesis of ADC dual-drug-linker
- Forms an important ADC dual-drug link assembly unit
- Related to peptide and derivatives
- Related to peptides for drug delivery
- Related to peptide linkers
- Related to cancer targeted therapy
- Related to ADC linker