Learn More
Medchemexpress LLC Fomc-Gly-Gly-Phe-Gly-OH | 1817857-75-2 | 99.7% | 558.58 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP4192100MG
Encompass_Preferred
Fomc-Gly-Gly-Phe-Gly-OH (compound D5) is an intermediate chemical utilized in the synthesis of Antibody-Drug Conjugate (ADC) dual-drug linkers. It serves as a precursor to Fomc-Gly-Gly-Phe-Gly-OH synthetic intermediate GGFGE, which is a crucial component in assembling ADC dual-drug link units. This product is intended for research use only.
  • Intermediate for ADC dual-drug-linker synthesis.
  • Forms an important ADC dual-drug link assembly unit.
  • High purity at 99.66%.
  • Available in various quantities (mg, g).
  • Molecular weight: 558.58.
  • Clearly defined CAS no. 1817857-75-2 for identification.
Catalog No. 50-003-96644