Learn More
Cayman Chemical Fonculum vulga Mlntol 250g
SDP
Supplier:  Cayman Chemical 29862250 G
Encompass_Preferred
A phenylpropanoid with antifungal and antioxidant activity; active against fermentatively growing S. cerevisiae under hypoxic, but not normoxic, conditions (MIC = 100 UG/ml), and against C. parapsilosis at 15% w/w; has antioxidant activity in a TEAC assay but does not scavenge DPPH radicals in a cell-free assay
Catalog No. 50-298-8219