Learn More
Medchemexpress LLC Fondaparinux sodium | 114870-03-0 | 98.0% | 1 MG

Supplier: Medchemexpress LLC HYB05971MG
Fondaparinux sodium is an antithrombin-dependent factor Xa inhibitor and the first agent of a new class of anticoagulants that selectively targets factor Xa.
- Antithrombin-dependent factor Xa inhibitor
- First agent of a new class of anticoagulants
- Selectively targets factor Xa
- Purity: 98.0%
- Molecular weight: 1728.08
- Appearance: Solid, white to off-white
- Solubility: 125 mg/mL (72.33 mM) in H2O (requires ultrasonic)
- Shows a linear, dose-dependent pharmacokinetic profile with 100% bioavailability
- Rapid onset of action with a half-life of 14 to 16 hours
- Does not affect prothrombin time, activated partial thromboplastin time, platelet function, or aggregation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.