Learn More
Medchemexpress LLC Fondaparinux sodium | 114870-03-0 | ≥98.0% | 1728.08 | C31H43N3Na10O49S8 | 10 MG

Supplier: Medchemexpress LLC HYB059710MG
Fondaparinux sodium is a synthetic pentasaccharide anticoagulant that selectively inhibits factor Xa through antithrombin binding. It is supplied as the sodium salt for research and analytical applications, and is commonly used as a selective biochemical reagent in coagulation and pharmacology studies.
- Antithrombin-dependent factor Xa inhibitor.
- Synthetic pentasaccharide sodium salt.
- Molecular weight 1728.08 with defined chemical formula.
- Purity ≥98.0% as tested.
- Supplied in a 10 MG quantity for research use.
- Store sealed at 4°C; in solvent keep at -80°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.