Learn More
Selleck Chemical LLC Fondaparinux Sodium (Org 31540) S3736-5mg
SDP
Supplier:  Selleck Chemical LLC S37365MG
Encompass_Preferred
Fondaparinux Sodium (Org 31540 Natural heparin pentasaccharide Fondaparin SR-90107A) is a synthetic glucopyranoside with antithrombotic activity It selectively binds to antithrombin III thereby potentiating the innate neutralization of activated factor X (Factor Xa) by antithrombin a synthetic inhibitor of Factor Xa
Catalog No. 50-217-3125