Learn More
STEMCELL Technologies Caspase-3/7 Activity Plate Reader Assay Kit, Green, Size: 200 Tests
SDP
Supplier:  STEMCELL Technologies 1000920
Encompass
Efficiently detect caspase-3/7 activity in cell lysates prepared from mammalian cells undergoing apoptosis by using the Caspase-3/7 Activity Plate Reader Assay Kit, Green. This reagent kit includes the fluorogenic substrate Z-DEVD-Rh 110-DVED-Z and an assay buffer. With its quick and simple staining protocol, this kit is well suited for high-throughput screening and compatible with standard fluorescent plate readers. Z-DEVD-Rh 110-DVED-Z has an excitation wavelength of 500 nm and a fluorescence emission wavelength of 522 nm. When added to apoptotic cells, Z-DEVD-Rh 110-DVED-Z is cleaved by activated caspase-3/7, producing a green fluorescent signal that can be detected by standard plate readers. Uses of caspase assays include quantifying caspase-3/7 activity in apoptotic cells and for screening caspase-3/7 inhibitors. Visit STEMCELL.com for more information and related caspase assays.
Catalog No. 50-225-0332