Learn More
Med Vet International Miltex Meisterhand Babcock Tissue Forceps, 9.5in, Jaw Width 14.7mm, German, Mh16-48
Supplier:  Med Vet International MILMH1648
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Babcock Tissue Forceps. Babcock Forceps are used to grasp delicate tissue in laser procedures. They are made from premium-grade German stainless steel. Specifications: 9.5in, Jaw Width 14.7mm, MH16-48
Catalog No. 50-283-1210