Learn More
Med Vet International Miltex Meisterhand Bonney Tissue Forceps, 6.75in, 1 X 2 Teeth, Serrated Tips, German, Mh6-148
Supplier:  Med Vet International MILMH6148
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Bonney Tissue Forceps. Bonney Tissue Forceps are thumb forceps used for grasping heavy tissue or bone. They are made from premium-grade German stainless steel. Specifications: 6.75in, 1 X 2 Teeth, Serrated Tips, MH6-148
Catalog No. 50-283-2342